(1R)-1-(3,4-Dichlorophenyl)propan-1-amine hydrochloride ≥97%

Supplier: Matrix Scientific

087240-500MG 087240-1G
102853-026EA 803.08 USD
102853-026 102853-028
(1R)-1-(3,4-Dichlorophenyl)propan-1-amine hydrochloride ≥97%
(1R)-1-(3,4-Dichlorophenyl)propan-1-amine hydrochloride

Hazard: Irritant

Formula: C₉H₁₂Cl₃N
MDL Number: MFCD12757847
CAS Number: 742107-61-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR