3-Cyano-4-phenylpyridine-2-carboxylic acid ≥97%
Supplier: Matrix Scientific
087239-500MG
087239-1G
102853-022EA
995.38
USD
102853-022
102853-024
3-Cyano-4-phenylpyridine-2-carboxylic acid ≥97%
3-Cyano-4-phenylpyridine-2-carboxylic acid
Hazard: Irritant
Formula:
C₁₃H₈N₂O₂ Melting Pt: 172 °C (dec.) |
MDL Number:
MFCD22573524 CAS Number: 133609-28-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...