4-Chloro-5-(trifluoromethyl)pyridine-2-carboxylic acid ≥97%
Supplier: Matrix Scientific
087238-500MG
087238-1G
102853-018EA
1240
USD
102853-018
102853-020
4-Chloro-5-(trifluoromethyl)pyridine-2-carboxylic acid ≥97%
4-Chloro-5-(trifluoromethyl)pyridine-2-carboxylic acid
Hazard: Irritant
Formula:
C₇H₃ClF₃NO₂ MW: 225.56 g/mol Melting Pt: 150 °C |
MDL Number:
MFCD18257442 CAS Number: 1211591-26-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...