(4-Chlorothiophen-2-yl)(4-fluorophenyl)methanone ≥97%

Supplier: Matrix Scientific

087237-500MG 087237-1G
102853-014EA 403.08 USD
102853-014 102853-016
(4-Chlorothiophen-2-yl)(4-fluorophenyl)methanone ≥97%
(4-Chlorothiophen-2-yl)(4-fluorophenyl)methanone

Hazard: Irritant

Formula: C₁₁H₆ClFOS
MDL Number: MFCD22573546
CAS Number: 207852-56-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR