2-Chloro-3-iodo-5-methyl-1,6-naphthyridine ≥97%

Supplier: Matrix Scientific

087230-500MG 087230-1G
102852-986EA 1192.31 USD
102852-986 102852-988
2-Chloro-3-iodo-5-methyl-1,6-naphthyridine ≥97%
2-Chloro-3-iodo-5-methyl-1,6-naphthyridine

Hazard: Irritant

Formula: C₉H₆ClIN₂
Melting Pt: 172…174 °C
MDL Number: MFCD22573535
CAS Number: 1407521-91-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR