4-Bromo-N-(1,3-thiazol-2-yl)benzene-1-sulfonamide ≥97%

Supplier: Matrix Scientific

087228-500MG 087228-1G
102852-978EA 403.08 USD
102852-978 102852-980
4-Bromo-N-(1,3-thiazol-2-yl)benzene-1-sulfonamide ≥97%
4-Bromo-N-(1,3-thiazol-2-yl)benzene-1-sulfonamide

Hazard: Irritant

Formula: C₉H₇BrN₂O₂S₂
MW: 319.21 g/mol
MDL Number: MFCD00592646
CAS Number: 331972-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR