4-Bromo-N-(1,3,4-thiadiazol-2-yl)benzene-1-sulfonamide ≥97%

Supplier: Matrix Scientific

087226-500MG 087226-1G
102852-972EA 600 USD
102852-972 102852-974
4-Bromo-N-(1,3,4-thiadiazol-2-yl)benzene-1-sulfonamide ≥97%
4-Bromo-N-(1,3,4-thiadiazol-2-yl)benzene-1-sulfonamide

Hazard: Irritant

Formula: C₈H₆BrN₃O₂S₂
MW: 320.19 g/mol
MDL Number: MFCD02861948
CAS Number: 349614-28-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR