4-Bromo-1-ethyl-2-fluorobenzene ≥95%

Supplier: Matrix Scientific

087221-500MG 087221-1G
102852-954EA 656.92 USD
102852-954 102852-956
4-Bromo-1-ethyl-2-fluorobenzene ≥95%
4-Bromo-1-ethyl-2-fluorobenzene

Hazard: Irritant

Formula: C₈H₈BrF
MDL Number: MFCD16619427
CAS Number: 627463-18-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR