3-(2-Bromo-4,5-dimethoxyphenyl)propanenitrile ≥97%

Supplier: Matrix Scientific

087220-500MG 087220-1G
102852-950EA 298.46 USD
102852-950 102852-952
3-(2-Bromo-4,5-dimethoxyphenyl)propanenitrile ≥97%
3-(2-Bromo-4,5-dimethoxyphenyl)propanenitrile

Hazard: Irritant

Formula: C₁₁H₁₂BrNO₂
MDL Number: MFCD00100650
CAS Number: 35249-62-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR