8-Bromoquinolin-2(1H)-one ≥97%

Supplier: Matrix Scientific

087219-500MG 087219-1G
102852-946EA 193.85 USD
102852-946 102852-948
8-Bromoquinolin-2(1H)-one ≥97%
8-Bromoquinolin-2(1H)-one

Hazard: Irritant

Formula: C₉H₆BrNO
MDL Number: MFCD08741409
CAS Number: 67805-67-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR