Benzyl-4-[methoxy(methyl)carbamoyl]piperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

087216-500MG 087216-1G
102852-932EA 403.08 USD
102852-932 102852-934
Benzyl-4-[methoxy(methyl)carbamoyl]piperidine-1-carboxylate ≥95%
Benzyl-4-[methoxy(methyl)carbamoyl]piperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₆H₂₂N₂O₄
MDL Number: MFCD07370044
CAS Number: 148148-48-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR