1-(5-Amino-1-methyl-1H-pyrazol-4-yl)ethan-1-one ≥97%

Supplier: Matrix Scientific

087213-500MG 087213-1G
102852-924EA 803.08 USD
102852-924 102852-926
1-(5-Amino-1-methyl-1H-pyrazol-4-yl)ethan-1-one ≥97%
1-(5-Amino-1-methyl-1H-pyrazol-4-yl)ethan-1-one

Hazard: Irritant

Formula: C₆H₉N₃O
MDL Number: MFCD19213343
CAS Number: 856860-17-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR