4-Amino-6-methyl-2-(methylsulfanyl)pyrimidine-5-carbonitrile ≥97%

Supplier: Matrix Scientific

087212-500MG 087212-1G
102852-920EA 253.85 USD
102852-920 102852-922
4-Amino-6-methyl-2-(methylsulfanyl)pyrimidine-5-carbonitrile ≥97%
4-Amino-6-methyl-2-(methylsulfanyl)pyrimidine-5-carbonitrile

Hazard: Irritant

Formula: C₇H₈N₄S
MW: 180.23 g/mol
Melting Pt: 237…239 °C
MDL Number: MFCD20693969
CAS Number: 89853-27-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR