4-Amino-2-methoxy-6-methylpyrimidine-5-carbonitrile ≥97%

Supplier: Matrix Scientific

087211-500MG 087211-1G
102852-916EA 403.08 USD
102852-916 102852-918
4-Amino-2-methoxy-6-methylpyrimidine-5-carbonitrile ≥97%
4-Amino-2-methoxy-6-methylpyrimidine-5-carbonitrile

Hazard: Irritant

Formula: C₇H₈N₄O
Melting Pt: 240…242 °C
MDL Number: MFCD20718650
CAS Number: 209849-42-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR