1-Acetylcyclopropane-1-carboxylic acid ≥95%

Supplier: Matrix Scientific

087208-500MG 087208-1G
102852-904EA 600 USD
102852-904 102852-906
1-Acetylcyclopropane-1-carboxylic acid ≥95%
1-Acetylcyclopropane-1-carboxylic acid

Hazard: Irritant

Formula: C₆H₈O₃
MDL Number: MFCD11226172
CAS Number: 56172-71-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR