3-Bromo-2,6-difluoropyridine ≥98%

Supplier: Matrix Scientific

087201-1G 087201-5G
102852-896EA 44.62 USD
102852-896 102852-898
3-Bromo-2,6-difluoropyridine ≥98%
3-Bromo-2,6-difluoropyridine

Hazard: Irritant

Formula: C₅H₂BrF₂N
MDL Number: MFCD11044274
CAS Number: 80392-79-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR