3-Bromo-6-chloro-2-fluoropyridine ≥98%

Supplier: Matrix Scientific

087200-1G 087200-5G
102852-892EA 80 USD
102852-892 102852-894
3-Bromo-6-chloro-2-fluoropyridine ≥98%
3-Bromo-6-chloro-2-fluoropyridine

Hazard: Irritant

Formula: C₅H₂BrClFN
MDL Number: MFCD06798230
CAS Number: 885952-18-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR