2-Bromo-3-(trifluoromethyl)benzoic acid ≥98%

Supplier: Matrix Scientific

087191-1G 087191-5G
102852-888EA 164.62 USD
102852-888 102852-890
2-Bromo-3-(trifluoromethyl)benzoic acid ≥98%
2-Bromo-3-(trifluoromethyl)benzoic acid

Hazard: Irritant

Formula: C₈H₄BrF₃O₂
MW: 269.02 g/mol
Melting Pt: 143-145°C
MDL Number: MFCD08532493
CAS Number: 177420-63-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR