(3-Bromo-5-(trifluoromethyl)phenyl)methanol ≥98%

Supplier: Matrix Scientific

Synonyms: 3-Bromo-5-(trifluoromethyl)benzyl alcohol

087183-1G 087183-5G
102852-878EA 36.92 USD
102852-878 102852-880
(3-Bromo-5-(trifluoromethyl)phenyl)methanol ≥98%
(3-Bromo-5-(trifluoromethyl)phenyl)methanol

Hazard: Irritant

Formula: C₈H₆BrF₃O
MDL Number: MFCD09835207
CAS Number: 172023-97-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR