6-Oxo-6-(9-Phenanthryl)hexanoic acid ≥97%

Supplier: Matrix Scientific

086987-1G 086987-2G 086987-5G
102851-818EA 476.92 USD
102851-818 102851-820 102851-822
6-Oxo-6-(9-Phenanthryl)hexanoic acid ≥97%
6-Oxo-6-(9-Phenanthryl)hexanoic acid

Hazard: Irritant

Formula: C₂₀H₁₈O₃
MDL Number: MFCD01320095
CAS Number: 898766-05-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR