3-(3,4-Dimethoxyphenyl)-2-methyl-1-propene ≥97%
Supplier: Matrix Scientific
086260-1G
086260-5G
102847-632EA
380
USD
102847-632
102847-636
3-(3,4-Dimethoxyphenyl)-2-methyl-1-propene ≥97%
3-(3,4-Dimethoxyphenyl)-2-methyl-1-propene
Hazard: Irritant
Formula:
C₁₂H₁₆O₂ |
MDL Number:
MFCD07775001 CAS Number: 155583-45-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...