3-Chloro-5-(trifluoromethyl)benzoic acid ≥95%
Supplier: Matrix Scientific
009469-1G
009469-5G
102832-804EA
29.23
USD
102832-804
102832-806
3-Chloro-5-(trifluoromethyl)benzoic acid ≥95%
3-Chloro-5-(trifluoromethyl)benzoic acid
Hazard: Irritant
Formula:
C₈H₄ClF₃O₂ Melting Pt: 98…101 °C |
MDL Number:
MFCD04115846 CAS Number: 53985-49-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...