(1,3-Dioxolan-2-yl)acetic acid ethyl ester
Supplier: Matrix Scientific
087156-1G
087156-2G
087156-5G
102852-832EA
572.31
USD
102852-832
102852-834
102852-836
(1,3-Dioxolan-2-yl)acetic acid ethyl ester
(1,3-Dioxolan-2-yl)acetic acid ethyl ester
Hazard: Irritant
Formula:
C₇H₁₂O₄ |
MDL Number:
MFCD18433503 CAS Number: 60234-78-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...