4-(5,5-Dimethyl-1,3-dioxan-2-yl)-2'-fluorobutyrophenone ≥97%

Supplier: Matrix Scientific

087152-1G 087152-2G 087152-5G
102852-808EA 580 USD
102852-808 102852-810 102852-812
4-(5,5-Dimethyl-1,3-dioxan-2-yl)-2'-fluorobutyrophenone ≥97%
4-(5,5-Dimethyl-1,3-dioxan-2-yl)-2'-fluorobutyrophenone

Hazard: Irritant

Formula: C₁₆H₂₁FO₃
Melting Pt: 60…61 °C
MDL Number: MFCD03844241
CAS Number: 898786-04-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR