4'-Chloro-3-(1,3-dioxan-2-yl)propiophenone ≥97%

Supplier: Matrix Scientific

087151-1G 087151-2G 087151-5G
102852-802EA 592.31 USD
102852-802 102852-804 102852-806
4'-Chloro-3-(1,3-dioxan-2-yl)propiophenone ≥97%
4'-Chloro-3-(1,3-dioxan-2-yl)propiophenone

Hazard: Irritant

Formula: C₁₃H₁₅ClO₃
MDL Number: MFCD02261796
CAS Number: 724708-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR