3'-Chloro-3-(1,3-dioxan-2-yl)propiophenone ≥97%
Supplier: Matrix Scientific
087150-1G
087150-2G
087150-5G
102852-796EA
590.77
USD
102852-796
102852-798
102852-800
3'-Chloro-3-(1,3-dioxan-2-yl)propiophenone ≥97%
3'-Chloro-3-(1,3-dioxan-2-yl)propiophenone
Hazard: Irritant
Formula:
C₁₃H₁₅ClO₃ |
MDL Number:
MFCD02261795 CAS Number: 898785-84-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...