2'-Chloro-3-(1,3-dioxan-2-yl)propiophenone ≥97%

Supplier: Matrix Scientific

087149-1G 087149-2G 087149-5G
102852-790EA 580 USD
102852-790 102852-792 102852-794
2'-Chloro-3-(1,3-dioxan-2-yl)propiophenone ≥97%
2'-Chloro-3-(1,3-dioxan-2-yl)propiophenone

Hazard: Irritant

Formula: C₁₃H₁₅ClO₃
MDL Number: MFCD02261794
CAS Number: 898785-82-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR