1-Bromo-3-(1,3-dioxolan-2-yl)-4-methoxybenzene ≥97%

Supplier: Matrix Scientific

087148-1G 087148-2G 087148-5G
102852-784EA 489.23 USD
102852-784 102852-786 102852-788
1-Bromo-3-(1,3-dioxolan-2-yl)-4-methoxybenzene ≥97%
1-Bromo-3-(1,3-dioxolan-2-yl)-4-methoxybenzene

Hazard: Irritant

Formula: C₁₀H₁₁BrO₃
MDL Number: MFCD06208940
CAS Number: 156603-10-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR