1-Bromo-4-(1,3-dioxolan-2-yl)-3-fluorobenzene ≥97%

Supplier: Matrix Scientific

087146-1G 087146-2G 087146-5G
102852-772EA 260 USD
102852-772 102852-774 102852-776
1-Bromo-4-(1,3-dioxolan-2-yl)-3-fluorobenzene ≥97%
1-Bromo-4-(1,3-dioxolan-2-yl)-3-fluorobenzene

Hazard: Irritant

Formula: C₉H₈BrFO₂
Melting Pt: 42…45 °C
MDL Number: MFCD06209216
CAS Number: 248270-23-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR