3-(4-Chlorophenyl)propionaldehyde ≥97%

Supplier: Matrix Scientific

Synonyms: 3-(4-Chlorophenyl)propanal

087140-1G 087140-2G 087140-5G
102852-736EA 769.23 USD
102852-736 102852-738 102852-740
3-(4-Chlorophenyl)propionaldehyde ≥97%
3-(4-Chlorophenyl)propionaldehyde

Hazard: Irritant

Formula: C₉H₉ClO
MW: 168.62 g/mol
MDL Number: MFCD07772912
CAS Number: 75677-02-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR