3-(3-Methylphenyl)propionaldehyde ≥97%
Supplier: Matrix Scientific
087138-1G
087138-2G
087138-5G
102852-724EA
769.23
USD
102852-724
102852-726
102852-728
3-(3-Methylphenyl)propionaldehyde ≥97%
3-(3-Methylphenyl)propionaldehyde
Hazard: Irritant
Formula:
C₁₀H₁₂O |
MDL Number:
MFCD07772907 CAS Number: 95416-60-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...