3-(4-Methylphenyl)propionaldehyde ≥97%

Supplier: Matrix Scientific

087137-1G 087137-2G 087137-5G
102852-718EA 769.23 USD
102852-718 102852-720 102852-722
3-(4-Methylphenyl)propionaldehyde ≥97%
3-(4-Methylphenyl)propionaldehyde

Hazard: Irritant/Stored under Argon

Formula: C₁₀H₁₂O
Boiling Pt: 223 °C
Density: 0.9928/18°C
MDL Number: MFCD04112993
CAS Number: 5406-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR