3-(3-Methoxyphenyl)propionaldehyde ≥97%

Supplier: Matrix Scientific

087135-1G 087135-2G 087135-5G
102852-706EA 769.23 USD
102852-706 102852-708 102852-710
3-(3-Methoxyphenyl)propionaldehyde ≥97%
3-(3-Methoxyphenyl)propionaldehyde

Hazard: Irritant

Formula: C₁₀H₁₂O₂
MDL Number: MFCD07772909
CAS Number: 40138-66-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR