3-(3-Chlorophenyl)propionaldehyde ≥97%

Supplier: Matrix Scientific

087134-1G 087134-2G 087134-5G
102852-700EA 769.23 USD
102852-700 102852-702 102852-704
3-(3-Chlorophenyl)propionaldehyde ≥97%
3-(3-Chlorophenyl)propionaldehyde

Hazard: Irritant

Formula: C₉H₉ClO
MW: 168.62 g/mol
MDL Number: MFCD07772911
CAS Number: 136415-83-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR