3-(4-Methoxyphenyl)propionaldehyde ≥97%

Supplier: Matrix Scientific

Synonyms: 3-(4-Methoxyphenyl)propanal

087132-1G 087132-2G 087132-5G
102852-688EA 490.77 USD
102852-688 102852-690 102852-692
3-(4-Methoxyphenyl)propionaldehyde ≥97%
3-(4-Methoxyphenyl)propionaldehyde

Hazard: Irritant

Formula: C₁₀H₁₂O₂
MW: 164.21 g/mol
MDL Number: MFCD00673205
CAS Number: 20401-88-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR