1-(1,3-Dioxolan-2-ylmethyl)-4-methylbenzene ≥97%

Supplier: Matrix Scientific

087131-1G 087131-2G 087131-5G
102852-682EA 490.77 USD
102852-682 102852-684 102852-686
1-(1,3-Dioxolan-2-ylmethyl)-4-methylbenzene ≥97%
1-(1,3-Dioxolan-2-ylmethyl)-4-methylbenzene

Hazard: Irritant

Formula: C₁₁H₁₄O₂
MDL Number: MFCD01924578
CAS Number: 4412-50-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR