2-Ethylbenzaldehyde ≥97%

Supplier: Matrix Scientific

087130-1G 087130-2G 087130-5G
102852-676EA 387.69 USD
102852-676 102852-678 102852-680
2-Ethylbenzaldehyde ≥97%
2-Ethylbenzaldehyde

Hazard: Irritant

Formula: C₉H₁₀O
MW: 134.1751 g/mol
MDL Number: MFCD02261766
CAS Number: 22927-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR