Ethyl-5-(2,5-dimethoxyphenyl)-5-oxovalerate ≥97%

Supplier: Matrix Scientific

087122-1G 087122-2G 087122-5G
102852-628EA 592.31 USD
102852-628 102852-630 102852-632
Ethyl-5-(2,5-dimethoxyphenyl)-5-oxovalerate ≥97%
Ethyl-5-(2,5-dimethoxyphenyl)-5-oxovalerate

Hazard: Irritant

Formula: C₁₅H₂₀O₅
MDL Number: MFCD02261474
CAS Number: 898758-29-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR