Ethyl-4-oxo-4-(4-trifluoromethylphenyl)butyrate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 4-oxo-4-(4-(trifluoromethyl)phenyl)butanoate

087121-1G 087121-2G 087121-5G
102852-622EA 592.31 USD
102852-622 102852-624 102852-626
Ethyl-4-oxo-4-(4-trifluoromethylphenyl)butyrate ≥97%
Ethyl-4-oxo-4-(4-trifluoromethylphenyl)butyrate

Hazard: Irritant

Formula: C₁₃H₁₃F₃O₃
MW: 274.24 g/mol
MDL Number: MFCD02261349
CAS Number: 155722-95-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR