Ethyl-4-oxo-4-(3-trifluoromethylphenyl)butyrate ≥95%

Supplier: Matrix Scientific

087120-1G 087120-2G 087120-5G
102852-616EA 592.31 USD
102852-616 102852-618 102852-620
Ethyl-4-oxo-4-(3-trifluoromethylphenyl)butyrate ≥95%
Ethyl-4-oxo-4-(3-trifluoromethylphenyl)butyrate

Hazard: Irritant

Formula: C₁₃H₁₃F₃O₃
MDL Number: MFCD02261344
CAS Number: 166312-68-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR