Ethyl-5-(4-nitrophenyl)-5-oxovalerate ≥97%

Supplier: Matrix Scientific

087119-1G 087119-2G 087119-5G
102852-610EA 592.31 USD
102852-610 102852-612 102852-614
Ethyl-5-(4-nitrophenyl)-5-oxovalerate ≥97%
Ethyl-5-(4-nitrophenyl)-5-oxovalerate

Hazard: Irritant

Formula: C₁₃H₁₅NO₅
MDL Number: MFCD02261335
CAS Number: 898777-59-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR