Ethyl-4-(4-nitrophenyl)-4-oxobutyrate ≥97%

Supplier: Matrix Scientific

087118-1G 087118-2G 087118-5G
102852-604EA 758.46 USD
102852-604 102852-606 102852-608
Ethyl-4-(4-nitrophenyl)-4-oxobutyrate ≥97%
Ethyl-4-(4-nitrophenyl)-4-oxobutyrate

Hazard: Irritant

Formula: C₁₂H₁₃NO₅
MW: 251.24 g/mol
MDL Number: MFCD02261334
CAS Number: 15118-70-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR