Ethyl-8-(4-iodophenyl)-8-oxooctanoate ≥97%

Supplier: Matrix Scientific

087117-1G 087117-2G 087117-5G
102852-598EA 490.77 USD
102852-598 102852-600 102852-602
Ethyl-8-(4-iodophenyl)-8-oxooctanoate ≥97%
Ethyl-8-(4-iodophenyl)-8-oxooctanoate

Hazard: Irritant

Formula: C₁₆H₂₁IO₃
MDL Number: MFCD02261323
CAS Number: 898777-48-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR