Ethyl-3-acetylbenzoate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 3-acetylbenzoate

087115-1G 087115-2G 087115-5G
102852-586EA 184.62 USD
102852-586 102852-588 102852-590
Ethyl-3-acetylbenzoate ≥97%
Ethyl-3-acetylbenzoate

Hazard: Irritant

Formula: C₁₁H₁₂O₃
MDL Number: MFCD02261283
CAS Number: 37847-24-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR