Ethyl-4-(2-oxopropyl)benzoate ≥97%

Supplier: Matrix Scientific

087113-1G 087113-2G 087113-5G
102852-574EA 735.38 USD
102852-574 102852-576 102852-578
Ethyl-4-(2-oxopropyl)benzoate ≥97%
Ethyl-4-(2-oxopropyl)benzoate

Hazard: Irritant

Formula: C₁₂H₁₄O₃
MDL Number: MFCD02261282
CAS Number: 73013-51-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR