Ethyl-3-(2-oxopropyl)benzoate ≥97%

Supplier: Matrix Scientific

087112-1G 087112-2G 087112-5G
102852-568EA 761.54 USD
102852-568 102852-570 102852-572
Ethyl-3-(2-oxopropyl)benzoate ≥97%
Ethyl-3-(2-oxopropyl)benzoate

Hazard: Irritant

Formula: C₁₂H₁₄O₃
MDL Number: MFCD02261281
CAS Number: 73013-49-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR