Ethyl-3-propionylbenzoate ≥97%

Supplier: Matrix Scientific

087110-1G 087110-2G 087110-5G
102852-556EA 490.77 USD
102852-556 102852-558 102852-560
Ethyl-3-propionylbenzoate ≥97%
Ethyl-3-propionylbenzoate

Hazard: Irritant

Formula: C₁₂H₁₄O₃
MDL Number: MFCD02261278
CAS Number: 898776-68-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR