Ethyl-4-(1-oxo-2-phenylethyl)benzoate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 4-(2-phenylacetyl)benzoate

087107-1G 087107-2G 087107-5G
102852-538EA 490.77 USD
102852-538 102852-540 102852-542
Ethyl-4-(1-oxo-2-phenylethyl)benzoate ≥97%
Ethyl-4-(1-oxo-2-phenylethyl)benzoate

Hazard: Irritant

Formula: C₁₇H₁₆O₃
MW: 268.32 g/mol
MDL Number: MFCD07700159
CAS Number: 898776-56-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR