Ethyl-4-oxo-4-(4-pyridyl)butyrate ≥97%

Supplier: Matrix Scientific

087105-1G 087105-2G 087105-5G
102852-526EA 490.77 USD
102852-526 102852-528 102852-530
Ethyl-4-oxo-4-(4-pyridyl)butyrate ≥97%
Ethyl-4-oxo-4-(4-pyridyl)butyrate

Hazard: Irritant

Formula: C₁₁H₁₃NO₃
MDL Number: MFCD01320358
CAS Number: 25370-46-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR