Ethyl-4-oxo-4-(3-pyridyl)butyrate ≥97%
Supplier: Matrix Scientific
087104-1G
087104-2G
087104-5G
102852-520EA
607.69
USD
102852-520
102852-522
102852-524
Ethyl-4-oxo-4-(3-pyridyl)butyrate ≥97%
Ethyl-4-oxo-4-(3-pyridyl)butyrate
Hazard: Irritant
Formula:
C₁₁H₁₃NO₃ |
MDL Number:
MFCD01320356 CAS Number: 59086-27-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...